Learn More
Epredia™ Richard-Allan Scientific™ Saccomanno Fluid
Catalog No. p-4530214
Click to view available options
Quantity:
60 mL
1 gal.
This item is not returnable. View return policy
This item is not returnable. View return policy
Store specimens for weeks with Epredia™ Richard-Allan Scientific™ Saccomanno Fluid, which acts as an excellent general cytological fixative.
  • Prevents air drying artifacts
  • For upper respiratory tract cytology, most notably sputum
  • Each prefilled container is labeled with a do not drink warning
  • Clear containers and tinted solution make specimen identification easy
  • Ready-to-use cytology fixative is an alcohol and polyethylene glycol (Carbowax™) solution
Specifications
Product Type Fixative
Quantity 60 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.