Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Signature Series™ Pen-Fix™

Catalog No. 22050316 Shop All Epredia Products
Click to view available options
Quantity:
5 gal.
4 x 5 L
4 x 1 gal.
This item is not returnable. View return policy

Catalog No. 22-050-316

Supplier: Epredia™ 6105

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.

  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative

Specifications

Quantity 5 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.