Learn More
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution
Shop All Epredia Products
Click to view available options
Type:
Steiner-Steiner Silver Kit
Description
- Kit does not contain uranyl nitrate
- Background is stained yellow to brown
- Bacteria, Donovan bodies and fungi all stain black
- Kit formulation and detailed procedures provide consistent and reproducible results
Specifications
Specifications
Type | Steiner-Steiner Silver Kit |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov