Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
Catalog No. 22050272 Shop All Epredia Products
Click to view available options
Type:
Epredia Three Step Stain Kit
Fixative
Solution A
Solution B
This item is not returnable. View return policy
This item is not returnable. View return policy
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
  • Stain can be used to detect H. Pylori.
  • Set contains easy-to-follow instructions
  • Product is available in one-gallon containers to facilitate use on automated strainers
  • Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)
Specifications
Type Epredia Three Step Stain Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.