Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rictor Antibody (7B3), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP151645APC
Description
Rictor Monoclonal specifically detects Rictor in Human, Mouse, Primate samples. It is validated for Flow Cytometry.Specifications
Rictor | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
RICTOR | |
Purified recombinant fragment of human RICTOR expressed in E. coli. [UniProt# Q6R327] | |
0.1 mL | |
Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway | |
253260 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
7B3 | |
Flow Cytometry | |
AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Mouse, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction