Learn More
Rictor Antibody (7B3), DyLight 550, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP151645R
Catalog No. NBP151645R
Only null left
Add to Cart
Description
Description
Rictor Monoclonal specifically detects Rictor in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Rictor
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
RICTOR
Purified recombinant fragment of human RICTOR expressed in E. coli. [UniProt# Q6R327]
0.1 mL
Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway
253260
Store at 4C in the dark.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
7B3
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human, Mouse, Primate
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only