Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rictor Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100611AF532
Description
Rictor Polyclonal specifically detects Rictor in Human samples. It is validated for Western Blot.Specifications
Rictor | |
Polyclonal | |
Western Blot | |
AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
Rabbit | |
Affinity Purified | |
Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway | |
253260.0 | |
Human | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
RICTOR | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Rapamycin-Insensitive Companion of mTOR using the numbering given in TrEMBL entry Q6R327 (GeneID 253260). | |
0.1 mL | |
Primary | |
NB100-611 is specific for human Rictor protein. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction