Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rictor Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100611IR
Description
Rictor Polyclonal specifically detects Rictor in Human samples. It is validated for Western Blot.Specifications
Rictor | |
Polyclonal | |
Western Blot | |
AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
NB100-611 is specific for human Rictor protein. | |
Store at 4C in the dark. |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RICTOR | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Rapamycin-Insensitive Companion of mTOR using the numbering given in TrEMBL entry Q6R327 (GeneID 253260). | |
100 μL | |
Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway | |
253260 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction