Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RICTOR Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518370
Description
This antibody is predicted to react with canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
Specifications
| RICTOR | |
| Polyclonal | |
| Unconjugated | |
| Rictor | |
| 4921505C17Rik; 6030405M08Rik; AVO3; AVO3 homolog; AW492497; D530039E11Rik; DKFZp686B11164; Fyb; FYN binding protein; hAVO3; Kiaa1999; mAVO3; MGC39830; PIA; pianissimo; protein pianissimo; Rapamycin-insensitive companion of mTOR; rictor; RPTOR independent companion of MTOR complex 2; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 253260 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6R327 | |
| Rictor | |
| Synthetic peptide sequence (KQPIVDTSAES) corresponding to the C-terminus amino acids of RICTOR (aa 1698-1708). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction