Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RICTOR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517451
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517451 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517451 Supplier Invitrogen™ Supplier No. PA517451
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RICTOR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Rictor
Gene Accession No. Q6QI06, Q6R327
Gene Alias 4921505C17Rik; 6030405M08Rik; AVO3; AVO3 homolog; AW492497; D530039E11Rik; DKFZp686B11164; Fyb; FYN binding protein; hAVO3; Kiaa1999; mAVO3; MGC39830; PIA; pianissimo; protein pianissimo; Rapamycin-insensitive companion of mTOR; rictor; RPTOR independent companion of MTOR complex 2; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3
Gene Symbols Rictor
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Lys1125 of human rictor.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253260, 310131, 78757
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.