Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RICTOR Recombinant Rabbit Monoclonal Antibody (1D5)
GREENER_CHOICE

Catalog No. PIMA545975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545975 Supplier Invitrogen™ Supplier No. MA545975
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Predicted band size: 192 kDa Positive Control: A549 cell lysate, SW480 cell lysate, A431, Hela. Subcellular Location: Cytosol, TORC2 complex. Sequence Similarities: 95.02% Mouse; 95.52% Rat. Tissue Specificity: Low tissue specificity.

RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RICTOR
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide
Gene Rictor
Gene Accession No. Q6R327
Gene Alias 4921505C17Rik; 6030405M08Rik; AVO3; AVO3 homolog; AW492497; D530039E11Rik; DKFZp686B11164; Fyb; FYN binding protein; hAVO3; Kiaa1999; mAVO3; MGC39830; PIA; pianissimo; protein pianissimo; Rapamycin-insensitive companion of mTOR; rictor; RPTOR independent companion of MTOR complex 2; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3
Gene Symbols Rictor
Host Species Rabbit
Immunogen Recombinant protein within human RICTOR aa 1459-1708.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253260
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.