Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products Ridgeline™ Full Brim Hard Hat, 4-Point Ratchet Suspension

Catalog No. p-7182076 Shop All Pyramex Safety Products Products
Click to view available options
Color:
Gray
White

Catalog No. 19-555-102

Supplier: Pyramex Safety Products HP54112

Only null left
Add to Cart
Add to Cart

Ridgeline™ Full Brim Hard Hat with 4-Point Ratchet Suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat.

  • Shell is constructed from ABS materials
  • Soft brow pad is replaceable
  • Replaceable suspension and headband also available

Specifications

Certifications/Compliance ANSI Z89.1
Color Gray
For Use With (Application) Construction
Material ABS
Product Type 4-point Ratchet Suspension
Size 6.5 to 8
Ventilated Not Vented
Head Style Full Brim
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.