Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rif1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001586H
Description
Rif1 Polyclonal antibody specifically detects Rif1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Rif1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp434D1026, DKFZp781N1478, FLJ10599, FLJ12870, RAP1 interacting factor homolog (yeast), Rap1-interacting factor 1 homolog, telomere-associated protein RIF1 | |
The immunogen recognized by this antibody maps to a region between residues 1425 and 1450 of human Rap1 Interacting Factor 1 using the numbering given in entry NP_060621.3 (GeneID 55183). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55183 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction