Learn More
RIF1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300568AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May have a role in sperm development and fertilization.Specifications
RIF1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5UIP0 | |
RIF1 | |
Between 1425 and 1450 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RIF1 | |
RAP1 interacting factor homolog, yeast, DKFZp434D1026, DKFZp781N1478, FLJ12870, rap1-interacting factor 1 homolog, telomere-associated protein RIF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55183 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.