Learn More
RIF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300567A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
May have a role in sperm development and fertilization.Specifications
| RIF1 | |
| Polyclonal | |
| Unconjugated | |
| RIF1 | |
| RAP1 interacting factor homolog, yeast, DKFZp434D1026, DKFZp781N1478, FLJ12870, rap1-interacting factor 1 homolog, telomere-associated protein RIF1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55183 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q5UIP0 | |
| RIF1 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.