Learn More
Rif1Rabbit anti-Mouse, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 600401871

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse Rif1 protein. A BLAST analysis was used to suggest reactivity with this protein from mouse based on 100% homology for the immunogen sequence. Expect cross reactivity with Rif1 from human, chimpanzee, rat and dog as only a single amino acid residue change (92% homology) is found for the immunogen sequence. Cross reactivity with Rif1 homologues from other sources has not been determined. Databases confirm that the immunogen sequence represents the C-terminal end of mouse Rif1 (534 aa 58.1 kDa), mouse telomere-associated protein RIF1 (2418 aa 265.7 kDa) and mouse Rap1-interacting factor 1 (2426 267.1 kDa).
May have a role in sperm development and fertilization.Specifications
Rif1 | |
Polyclonal | |
Unconjugated | |
RIF1 | |
5730435J01Rik; 6530403D07Rik; AU016181; AW549474; D2Ertd145e; DKFZp434D1026; DKFZp781N1478; FLJ12870; mRif1; Rap1 interacting factor 1 homolog (yeast); RAP1 interacting factor homolog; Rap1-interacting factor 1 homolog; replication timing regulatory factor 1; Rif1; Telomere-associated protein RIF1 | |
Rabbit | |
RUO | |
51869 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Q6PR54 | |
RIF1 | |
100 μg | |
Primary | |
Mouse | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.