Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifampin, 50 mg/ml in DMSO, sterile-filtered

Catalog No. AAJ67397XF
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67397XF 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67397XF Supplier Thermo Scientific Chemicals Supplier No. J67397XF
Add to Cart

CAS: 13292-46-1 | C43H58N4O12 | 822.953 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
MDL Number MFCD00151389
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rifampicin, 50 mg/ml in DMSO, sterile-filtered
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Type Rifampicin
Physical Form Liquid
Packaging Plastic Bottle
Quantity 1 mL
Chemical Name or Material Rifampin
Hazard Category H227-H335
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 236-312-0
RTECSNumber VJ7000000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.