Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rift Valley Fever Virus Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396516 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396516 0.025 mg
NBP241081 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396516 Supplier Novus Biologicals Supplier No. NBP2410810.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rift Valley Fever Virus Polyclonal specifically detects Rift Valley Fever Virus in Virus samples. It is validated for ELISA.

Specifications

Antigen Rift Valley Fever Virus
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias Glycoprotein, RVFV_sM_gp1
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide near the center of the RVF virus. The immunogen is located within amino acids 360 - 410 of Rift Valley Fever Virus. Amino Acid Squence: CFEHKGGYKGTMDSGQTKRE
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.