Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rift Valley Fever Virus Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241082
Description
Rift Valley Fever Virus Polyclonal specifically detects Rift Valley Fever Virus in Virus samples. It is validated for ELISA.Specifications
| Rift Valley Fever Virus | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| This RVF virus antibody was derived from a peptide sequence near the carboxy terminus of the polyprotein precursor translated from the M segment. It will therefore detect both the precursor and the Glycoprotein G2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA | |
| Glycoprotein, RVFV_sM_gp1 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of the RVF virus. The immunogen is located within amino acids 960 - 1010 of Rift Valley Fever Virus. Amino Acid Squence: FERGSLPQTRNDKTFAASK | |
| 0.1 mg | |
| Primary | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction