Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rift Valley Fever Virus Nucleocapsid Mouse anti-Virus, Clone: DE1.3.B9, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP3148580.5MG
This item is not returnable.
View return policy
Description
Rift Valley Fever Virus Nucleocapsid Monoclonal antibody specifically detects Rift Valley Fever Virus Nucleocapsid in Virus samples. It is validated for ELISASpecifications
Rift Valley Fever Virus Nucleocapsid | |
Monoclonal | |
1 mg/mL | |
ELISA | |
Fragment, NP, nucleocapsid, Nucleocapsid protein, Nucleocapsid proteinNucleoprotein, Nucleoprotein, RVFV_sS_gp2 | |
Rift Valley Fever Virus Nucleoprotein, aa2-245. | |
0.5 mg | |
Infections (Virus Bacteria and Parasites) | |
9538291 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Purified |
ELISA | |
DE1.3.B9 | |
Unconjugated | |
Phosphate Buffered Saline, pH 7.4, Filter Sterile | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Virus | |
Antibody | |
IgG1a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction