Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rift Valley Fever Virus Nucleocapsid Mouse anti-Virus, Clone: IA4.1.D5.B8, Novus Biologicals™

Mouse Monoclonal Antibody
$515.50
Specifications
Antigen | Rift Valley Fever Virus Nucleocapsid |
---|---|
Clone | IA4.1.D5.B8 |
Concentration | 1 mg/mL |
Dilution | ELISA |
Applications | ELISA |
Description
Rift Valley Fever Virus Nucleocapsid Monoclonal antibody specifically detects Rift Valley Fever Virus Nucleocapsid in Virus samples. It is validated for ELISASpecifications
Rift Valley Fever Virus Nucleocapsid | |
1 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Infections (Virus Bacteria and Parasites) | |
Phosphate Buffered Saline, pH 7.4, Filter Sterile | |
9538291 | |
IgG1 κ | |
Protein G purified | |
Antibody |
IA4.1.D5.B8 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
Fragment, NP, nucleocapsid, Nucleocapsid protein, Nucleocapsid proteinNucleoprotein, Nucleoprotein, RVFV_sS_gp2 | |
Rift Valley Fever Virus Nucleoprotein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title