Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Rift Valley Fever Virus Polymerase Antibody, PerCP, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241224PCP
Description
Rift Valley Fever Virus Polymerase Polyclonal antibody specifically detects Rift Valley Fever Virus Polymerase in Virus samples. It is validated for ELISASpecifications
| Rift Valley Fever Virus Polymerase | |
| Polyclonal | |
| ELISA | |
| EC=2.7.7.48, Replicase, RVFV_sL_gp1, Transcriptase | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of the RVF virus Polymerase. The immunogen is located within amino acids 370 - 420 of Rift Valley Fever Virus Polymerase. Amino Acid Squence: VKDRPDERNRYHRVHL | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 9538294 | |
| Store at 4C in the dark. | |
| IgG | 
| ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            