Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIG-1/DDX58 Monoclonal Antibody (3B10F5), CoraLite488, Proteintech
SDP

Catalog No. 502333074 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-3074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-3074 Supplier Proteintech Group Inc Supplier No. CL48867556100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RIG-1/DDX58 Monoclonal antibody specifically detects RIG-1/DDX58 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry

Specifications

Antigen RIG-1/DDX58
Applications Immunocytochemistry
Classification Monoclonal
Clone 3B10F5
Concentration 1 mg/mL
Conjugate CoraLite 488
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Accession No. O95786
Gene Alias 6430573D20Rik; C330021E21; DDX58; RIG-I; RIGI; RLR-1; SGMRT2
Host Species Mouse
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23586
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.