Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Rig Dog™ Waterproof Mechanics Gloves

Catalog No. 19085650 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-650 Large
19-085-649 Medium
19-085-651 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-085-650 Supplier Honeywell™ Supplier No. 42615BL/9L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provides the protection of Xtreme gloves and adds a water-resistant barrier to offer comfort for wet applications.

  • Highly breathable, designed with an emphasis on wearer comfort
  • Water-resistant barrier offers protection and comfort in wet applications
  • Inner layer offers moisture absorption and a quick dry function to quickly absorb sweat and keep hands dry
  • Enhanced, level 2 impact protection from ANSI/ISEA 138
  • Additional layers provide A6/F level cut resistance, and guard against abrasions
  • Ergonomic design and flex-grooves provide increased dexterity
  • Slip-on closure

Specifications

Certifications/Compliance ANSI/ISEA 138 Level 2
Coating Material Uncoated
Color Black, Blue
Cut Resistance Rating ANSI A6
Material TPR
Product Type Mechanic's Gloves
Size Large
For Use With (Application) Drilling, material handling, rigging, ship building
Model 42-615BL/9L
Packaging Type Individual Polybag
Series Rig Dog™ Waterproof
Thumb Style Reinforced
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.