Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Rig Dog™ Xtreme Gloves

Catalog No. 19085637 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Glove Cuff Style:
Hook and Loop
Plain
Size:
Medium
Large
X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Style Size
19-085-637 Hook and Loop Medium
19-085-640 Plain Medium
19-085-638 Hook and Loop Large
19-085-641 Plain Large
19-085-639 Hook and Loop X-Large
19-085-642 Plain X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-085-637 Supplier Honeywell™ Supplier No. 42622BY/8M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provide cut protection, impact protection, and comfort with an enhanced grip for handling oily surfaces.

  • Made to keep workers safe and in control when handling hazardous oily surfaces
  • Highly breathable, designed with an emphasis on wearer comfort
  • Inner layer offers moisture absorption and a quick dry function to quickly absorb sweat and keep hands dry
  • Enhanced, level 2 impact protection from ANSI/ISEA 138
  • Additional layers provide A6/F level cut resistance, guard against abrasions & provide an enhanced grip

Specifications

Certifications/Compliance ANSI/ISEA 138 Level 2
Coating Material TPR
Color Black, Red, Yellow
Cut Resistance Rating ANSI A6
Hand Specific
Material TPR
Product Type Mechanic's Gloves
Size Medium
For Use With (Application) Handling Hazardous, Oily Surfaces
Glove Cuff Style Hook and Loop
Model 42-622BY/8M
Packaging Type Individual Polybag
Series Rig Dog™ Xtreme
Thumb Style Reinforced
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.