Learn More
Description
- Made to keep workers safe and in control when handling hazardous oily surfaces
- Highly breathable, designed with an emphasis on wearer comfort
- Inner layer offers moisture absorption and a quick dry function to quickly absorb sweat and keep hands dry
- Enhanced, level 2 impact protection from ANSI/ISEA 138
- Additional layers provide A6/F level cut resistance, guard against abrasions & provide an enhanced grip
Specifications
Specifications
| Certifications/Compliance | ANSI/ISEA 138 Level 2 |
| Coating Material | TPR |
| Color | Black, Red, Yellow |
| Cut Resistance Rating | ANSI A6 |
| Hand | Specific |
| Material | TPR |
| Product Type | Mechanic's Gloves |
| Size | Medium |
| For Use With (Application) | Handling Hazardous, Oily Surfaces |
| Glove Cuff Style | Plain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.