Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Rig Dog™ Xtreme Gloves

Catalog No. p-7203073
Click to view available options
Glove Cuff Style:
Hook and Loop
Plain
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Provide cut protection, impact protection, and comfort with an enhanced grip for handling oily surfaces.

  • Made to keep workers safe and in control when handling hazardous oily surfaces
  • Highly breathable, designed with an emphasis on wearer comfort
  • Inner layer offers moisture absorption and a quick dry function to quickly absorb sweat and keep hands dry
  • Enhanced, level 2 impact protection from ANSI/ISEA 138
  • Additional layers provide A6/F level cut resistance, guard against abrasions & provide an enhanced grip
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/ISEA 138 Level 2
Coating Material TPR
Color Black, Red, Yellow
Cut Resistance Rating ANSI A6
Hand Specific
Material TPR
Product Type Mechanic's Gloves
Size Medium
For Use With (Application) Handling Hazardous, Oily Surfaces
Glove Cuff Style Hook and Loop
Model 42-622BY/8M
Packaging Type Individual Polybag
Series Rig Dog™ Xtreme
Thumb Style Reinforced
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.