Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RIG-I Recombinant Rabbit Monoclonal Antibody (JE60-05)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544830
This item is not returnable.
View return policy
Description
RIG-I Recombinant Monoclonal Antibody for Western Blot
Retinoic acid-inducible gene I, RIG-I is a pattern recognition receptor (PRR) involved in the recognition of viral dsRNA. Along with MDA5, RIG-I detects viral dsRNA and activates the innate immune response. Both MDA5 and RIG-I are RNA helicases and they perform overlapping as well as distinct roles. RIG-I is activated by dsRNAs without a 5'-triphosphate end and short dsRNAs, whereas MDA5 is activated by long dsRNAs. Once activated, both proteins signal through IPS-1 activating transcription factors NF-kappaB and IRF-3 (1) and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus (3), paromyxoviruses, Japanese encephalitis virus, and West Nile virus. MicroRNA-146a has been implicated in feedback inhibition of RIG-I-dependant antiviral response by negatively regulating RIG-I targets TRAF6, IRAK1, and IRAK2. Recent evidence has implicated RIG-I in the detection of cytosolic DNA through RNA polymerase III activity.
Specifications
RIG-I | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95786 | |
DDX58 | |
Recombinant protein within human DDX58 aa 250-450/925. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
JE60-05 | |
Unconjugated | |
DDX58 | |
6430573D20Rik; C330021E21; Ddx58; DEAD (Asp-Glu-Ala-Asp) box polypeptide 58; DEAD box protein 58; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; DEAD/H box polypeptide RIG-I; DEAD-box protein 58; DEXD/H-box helicase 58; probable ATP-dependent RNA helicase DDX58; Retinoic acid-inducible gene 1 protein; retinoic acid-inducible gene I protein; retinoic acid-inducible gene-I; RIG-1; RIGI; RIG-I; RIG-I-like receptor 1; RLR-1; RNA helicase RIG-I; SGMRT2 | |
Rabbit | |
Protein A | |
RUO | |
23586 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction