Learn More
BINDER™ Right Access Port (10 to 30 mm)
Click to view available options
Diameter (Metric) Inside:
30 mm
Description
- For use on right
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Description | Right position |
For Use With (Equipment) | KBF 240, KBF-S 240, KBF-720, KBF-S 720, KBF 1020, KBF 115 |
Material | Steel |
Type | Access Port |
Includes | Silicone plug |
Diameter (Metric) Inside | 30 mm |