Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RILP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107015
Description
RILP Polyclonal Antibody for SPR, Western Blot
The Rab interacting lysosomal protein (RILP) is lysosomal protein that interacts with RAB7, a small GTPase that controls transport to endocytic degradative compartments. It is thought that RILP is a downstream effector of RAB7 and both proteins act together in the regulation of late endocytic traffic. Overexpression of RILP caused enlarged lysosomes that are more centrally located in the cell suggesting that RILP may also be involved in the regulation of lysosomal morphology. Other evidence suggests that RILP may play a role in the biogenesis of multivesicular bodies.
Specifications
| RILP | |
| Polyclonal | |
| Unconjugated | |
| RILP | |
| AI835553; Gm857; PP10141; Rab interacting lysosomal protein; rab-interacting lysosomal protein; RILP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83547 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Surface Plasmon Resonance | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q96NA2 | |
| RILP | |
| Synthetic peptide located within the following region: PPPPESKIQS FFGLWYRGKA ESSEDETSSP APSKLGGEEE AQPQSPAPDP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction