Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RILPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559512 Supplier Invitrogen™ Supplier No. PA559512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LELDRLRLER MDRIEKERKH QKELELVEDV WRGEAQDLLS QIAQLQEENK QLMTNLSHKD VNFSEEEFQK HEGMSERERQ VMKKLKEV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Plays a role in the regulation of cell shape and polarity. Plays a role in cellular protein transport, including protein transport away from primary cilia. Neuroprotective protein, which acts by sequestring GAPDH in the cytosol and prevent the apoptotic function of GAPDH in the nucleus. Competes with SIAH1 for binding GAPDH. Does not regulate lysosomal morphology and distribution (PubMed:14668488). Binds to RAB10 following LRRK2-mediated RAB10 phosphorylation which leads to inhibition of ciliogenesis (PubMed:30398148). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RILPL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RILPL1
Gene Accession No. Q5EBL4
Gene Alias 2900002H16Rik; 6330559I19Rik; GAPDH competitor of SIAH protein enhances life; GAPDH's competitor of SIAH1 protein enhances life; GOSPEL; MNCb-2440; Rab interacting lysosomal protein like 1; Rab interacting lysosomal protein-like 1; rab-interacting lysosomal-like protein 1; RGD1307973; Rilpl1; RILP-like protein 1; RLP1
Gene Symbols RILPL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RILPL1. Recombinant protein control fragment (Product #RP-97878).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353116
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.