Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RILPL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220162
Description
RILPL2 Polyclonal specifically detects RILPL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RILPL2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
FLJ30380, FLJ32372, MGC7036, p40phox-binding protein, Rab interacting lysosomal protein-like 2, Rab-interacting lysosomal protein-like 2, RILP-like protein 2, RLP2 | |
Rabbit | |
24 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Rat (80%), Bovine (84%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q969X0 | |
RILPL2 | |
Recombinant protein encompassing a sequence within the center region of human RILPL2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
196383 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction