Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RILPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534359
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534359 Supplier Invitrogen™ Supplier No. PA534359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is A549 cell lysate. PA5-34359 can be used with blocking peptide PEP-1402.

Does not regulate lysosomal morphology and distribution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RILPL2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Rilpl2
Gene Accession No. Q969X0
Gene Alias p40phox-binding protein; Rab interacting lysosomal protein like 2; Rab interacting lysosomal protein-like 2; rab-interacting lysosomal protein-like 2; Rab-interacting lysosomal-like protein 2; Rilpl2; RILP-like protein 2; RLP2
Gene Symbols Rilpl2
Host Species Rabbit
Immunogen An 16 amino acid synthetic peptide near the center of human RLP2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196383
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.