Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RILPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583047 Supplier Invitrogen™ Supplier No. PA583047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EERDEVGPEG ALGKSPFQLT AEDVYDISYL LGRELMALGS DPRVTQLQFK VVRVLEMLEA LVNEGSLALE ELKMERDHLR KEVEGLRRQS PPASGEVNLG PNKMVVDLTD PNRPRFTLQE LRDVLQERNK LK Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 81%.

Does not regulate lysosomal morphology and distribution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RILPL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rilpl2
Gene Accession No. Q969X0
Gene Alias p40phox-binding protein; Rab interacting lysosomal protein like 2; Rab interacting lysosomal protein-like 2; rab-interacting lysosomal protein-like 2; Rab-interacting lysosomal-like protein 2; Rilpl2; RILP-like protein 2; RLP2
Gene Symbols Rilpl2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RILPL2. Recombinant protein control fragment (Product #RP-89723).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196383
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.