Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520522
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520522 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520522 Supplier Invitrogen™ Supplier No. PA520522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20522 can be used with blocking peptide PEP-0642.

Rab3-interacting molecules (RIMs) are synaptic proteins necessary for neuronal transmission and plasticity. Rim1 and Rim2 proteins are expressed in overlapping but distinct patterns throughout the brain. While the ablation of either gene was not lethal in mice, the deletion of both resulted in postnatal mortality. This lethality is due to a defect in neurotransmitter release; synapses without RIM proteins can still release neurotransmitters but are unable to do so in response to normal Ca2+ triggers. Like Rim1, Rim2 is thought to be an effector protein for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIM2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Rims2
Gene Accession No. Q9EQZ7, Q9JIS1, Q9UQ26
Gene Alias 2810036I15Rik; AW048769; KIAA0751; mKIAA0751; Nim2; non-small cell lung cancer RimL3a protein; non-small cell lung cancer RimL3c protein; nuclear protein; OBOE; Rab3 interacting protein 1; Rab3 interacting protein 2; RAB3 interacting protein 3; Rab-3-interacting molecule 2; rab3-interacting molecule 2; Rab3-interacting protein; rab-3-interacting protein 2; rab3-interacting protein 2; Rab-3-interacting protein 3; Rab3ip2; RAB3IP3; regulating synaptic membrane exocytosis 2; regulating synaptic membrane exocytosis protein 2; RIM 2; Rim2; RIM2 (CT); Rim2(+40A); Rim2(+44A); Rim2(+4A); Rims2; Serg2; synaptic exocytosis regulator 2; synaptotagmin 3, related sequence; Syt3-rs
Gene Symbols Rims2
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of the human Rim2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116838, 116839, 9699
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.