Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RIM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565937
Description
Immunogen sequence: MVAIVGLTQWS KSTLQLPQPE GATKKLRSNI RR Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Rab3-interacting molecules (RIMs) are synaptic proteins are synaptic proteins necessary for neural transmission and plasticity. While both Rim1 and Rim 2 are thought to be effector proteins for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner, less is known of Rim3. Expression of Rim3 in PC12 cells induced a significant increase in calcium-triggered exocytosis, with no appreciable change in the baseline release, suggesting that it plays a role in the regulation of exocytosis. Rim3 protein localizes primarily to neuronal dendrites and the postsynaptic densities, as opposed to Rim1 which is found in presynapse locations, indicating that Rim3 may contribute to synapse transmission and plasticity.
Specifications
RIM3 | |
Polyclonal | |
Unconjugated | |
RIMS3 | |
A730060M23Rik; KIAA0237; mKIAA0237; Nim2; NIM3; Rab-3 interacting molecule 3; rab-3-interacting molecule 3; regulating synaptic membrane exocytosis 3; regulating synaptic membrane exocytosis protein 3; RIM 3; RIM3; RIM3 gamma; Rims3; RP4-739H11.2; si:ch211-239d6.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9783 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJD0 | |
RIMS3 | |
Recombinant Human RIM3. Recombinant protein control fragment (Product #RP-106619). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction