Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RIMBP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560614
Description
Immunogen sequence: RVDNITQISA QLSWLPTNSN YSHVIFLNEE EFDIVKAARY KYQFFNLRPN MAYKVKVLAK PHQMPWQLPL EQREKKEAFV EFSTLP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
The presynaptic active zone mediates synaptic vesicle exocytosis at a synapse. RIM proteins are active zone scaffolding molecules that--among others--mediate vesicle priming and directly or indirectly interact with most other essential presynaptic proteins. RIMBP2 encodes RIMS-binding protein 2 belonging to RIMBP family. RIMBP2 is supposed to be localized at cell membrane especially synaptic plasma membrane. By interacting with RIMS1, RIMS2, CACNA1D and CACNA1B via the SH3 domains, RIMBP2 mediates synaptic transmission as bifunctional linker.
Specifications
RIMBP2 | |
Polyclonal | |
Unconjugated | |
Rimbp2 | |
A930033C01Rik; KIAA0318; mKIAA0318; PPP1R133; protein phosphatase 1, regulatory subunit 133; RBP2; RIM binding protein 2; RIMBP2; RIM-BP2; RIMS binding protein 2; RIMS-binding protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23504 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15034 | |
Rimbp2 | |
Recombinant protein corresponding to Human RIMBP2. Recombinant protein control fragment (Product #RP-97658). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction