Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rimcazole dihydrochloride
GREENER_CHOICE

Catalog No. 149710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-971-0 10 mg
14-975-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-971-0 Supplier Tocris Bioscience™ Supplier No. 1497/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

σ2 antagonist. Also DAT inhibitor

Antagonist at σ receptors (IC50 values are 1480 and 386 nM at σ1 and σ2 receptors respectively). Also binds to the dopamine transporter (IC50 = 57.6 nM) and inhibits dopamine uptake. Reduces the stimulatory effects of cocaine in vivo.

Chemical Identifiers

CAS 75859-03-9
Molecular Formula C21H29Cl2N3

Specifications

Quantity 10 mg
Formula Weight 394.39
Purity >98%
Molecular Weight (g/mol) 394.384
Chemical Name or Material Rimcazole dihydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.