Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RIMKLA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101852
Description
Antibody detects endogenous levels of total RIMKA.
Catalyzes the synthesis of N-acetyl-L-aspartyl-L-glutamate (NAAG) and N-acetyl-L-aspartyl-L-glutamyl-L-glutamate.
Specifications
RIMKLA | |
Polyclonal | |
Unconjugated | |
RIMKLA | |
B930030J24; Fam80a; family with sequence similarity 80, member A; NAAG synthetase A; NAAGS; NAAGS-II; N-acetylaspartylglutamate synthase A; N-acetylaspartyl-glutamate synthetase A; N-acetylaspartylglutamate synthetase II; N-acetylaspartylglutamylglutamate synthase A; ribosomal modification protein rimK like family member A; ribosomal modification protein rimK-like family member A; ribosomal protein S6 modification-like protein A; Rimk; Rimkla; RP11-157D18.1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
194237, 284716 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6PFX8, Q8IXN7 | |
RIMKLA | |
A synthesized peptide derived from human RIMKLA(Accession Q8IXN7), corresponding to amino acid residues K111-G161. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction