Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIMKLB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599623
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599623 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599623 Supplier Invitrogen™ Supplier No. PA599623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FAM80B.

Catalyzes the synthesis of beta-citryl-L-glutamate and N-acetyl-L-aspartyl-L-glutamate. Beta-citryl-L-glutamate is synthesized more efficiently than N-acetyl-L-aspartyl-L-glutamate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIMKLB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RIMKLB
Gene Accession No. Q9ULI2
Gene Alias 4931417E21Rik; 4933426K21Rik; AA097693; AI325948; AU015629; beta-citrylglutamate synthase B; beta-citryl-glutamate synthase B; FAM80B; family with sequence similarity 80, member B; Kiaa1238; NAAG synthetase B; NAAGS; NAAGS-I; N-acetylaspartyl-glutamate synthetase; N-acetyl-aspartylglutamate synthetase B; N-acetyl-aspartyl-glutamate synthetase B; N-acetylaspartyl-glutamate synthetase B; RGD1560851; ribosomal modification protein rimK like family member B; ribosomal modification protein rimK-like family member B; ribosomal protein S6 modification-like protein B; Rimklb
Gene Symbols RIMKLB
Host Species Rabbit
Immunogen A synthesized peptide derived from human RIMKLB(Accession Q9ULI2), corresponding to amino acid residues H176-M226.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57494
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.