Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIMKLB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114104 Supplier Invitrogen™ Supplier No. PA5114104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FRAVVMDEMV LTVEQGNLGL RISGELISAY PQVVVVRVPT PWVQSDSDIT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 100%; Rat: 93%.

Catalyzes the synthesis of beta-citryl-L-glutamate and N-acetyl-L-aspartyl-L-glutamate. Beta-citryl-L-glutamate is synthesized more efficiently than N-acetyl-L-aspartyl-L-glutamate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIMKLB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RIMKLB
Gene Accession No. Q80WS1
Gene Alias 4931417E21Rik; 4933426K21Rik; AA097693; AI325948; AU015629; beta-citrylglutamate synthase B; beta-citryl-glutamate synthase B; FAM80B; family with sequence similarity 80, member B; Kiaa1238; NAAG synthetase B; NAAGS; NAAGS-I; N-acetylaspartyl-glutamate synthetase; N-acetyl-aspartylglutamate synthetase B; N-acetyl-aspartyl-glutamate synthetase B; N-acetylaspartyl-glutamate synthetase B; RGD1560851; ribosomal modification protein rimK like family member B; ribosomal modification protein rimK-like family member B; ribosomal protein S6 modification-like protein B; Rimklb
Gene Symbols RIMKLB
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Mouse Rimklb (aa 41-90).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108653
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.