Learn More
Eisco Labs Rimless Glass Test Tubes,
SCI_ED
Catalog No. S139053 Shop All Eisco Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
S139053 Autoclavable
1 options
Catalog No. S139053 Supplier Eisco Labs Supplier No. CH0718PPK24
Add to Cart
Add to Cart
Rimless Glass Test Tubes can be used for a variety of chemistry and biology activities.
  • Made of high quality, heavy duty Borosilicate 3.3 glass
  • Superior chemical durability and heat tolerance
  • Low temperature gradient can withstand direct flame, high temperatures, and thermal shocks
  • Flat, fire polished rims
  • Thin wall design
  • Autoclavable
Specifications
Capacity (Metric) 25 mL
Length (Metric) 20.32 mm
Material Borosilicate Glass (3.3)
Shape Cylindrical
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Product Type Test Tube