Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIMS2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241078
Description
RIMS2 Polyclonal specifically detects RIMS2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RIMS2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
RIMS2 | |
Antibody was raised against a 17 amino acid synthetic peptide near the center of human RIM2. The immunogen is located within amino acids 840 - 890 of RIM2. Amino Acid Squence: LPDRSDKNKRRTKTVKK | |
0.1 mg | |
Primary | |
RIM2 antibody is human specific. Multiple isoforms of RIM2 are known to exist. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
DKFZp781A0653, KIAA0751RIM2OBOERAB3 interacting protein 3, non-small cell lung cancer RimL3a protein, non-small cell lung cancer RimL3c protein, nuclear protein, rab3-interacting molecule 2, Rab-3-interacting molecule 2, Rab-3-interacting protein 3, RAB3IP3, regulating synaptic membrane exocytosis 2, regulating synaptic membrane exocytosis protein 2, RIM 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
9699 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction