Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110298 Supplier Invitrogen™ Supplier No. PA5110298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MESPGESGAG SPGAPSPSSF TTGHLAREKP AQDPLYDVPN ASGGQAGGPQ RPGRVVSLRE RLLLTRPVWL QLQANAAAAL HMLRTEPPGT FLVRKSNTRQ CQALCMRLPE ASGPSFVSSH YILESPGGVS LEGSELMFPD LVQLICAYCH TRDILLLPLQ LPRAIHHAAT HKELEAISHL GIEFWSSSLN IKAQRGPAGG PVLPQLKARS PQELDQGTGA ALCFFNPLFP GDLGPTKREK FKRSFKVRVS TETSSPLSPP AVPPPPVPVL PGAVPSQTER.

Rit and its neuron-specific homologue, Rin, define a recently discovered subfamily of Ras-related GTPases. Rit and Rin are membrane-associated in spite of the fact that they lack a CAAX box or similar C-terminal lipidation motif. Rit and Rin display 64% amino acid sequence identity and share a unique nine amino acid effector domain (DPTIEDAYK) that is 100% conserved between the murine and human proteins. Although the effector domain sequences of Rit and Rin are very similar to that of Ras, Rit and Rin have been shown to interact with the known Ras-binding proteins RalGDS, Rlf and AF-6, but not the Raf kinases, RIN1 or the p110 subunit of PI3 kinase. For this reason, it has been suggested that Rit and Rin may play important roles in the regulation of signaling pathways distinct from those controlled by Ras.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RIN1
Gene Accession No. P97680, Q921Q7
Gene Alias Ras and Rab interactor 1; ras inhibitor; ras inhibitor 1; ras inhibitor JC99; ras inhibitor RIN1; ras interaction/interference protein 1; RIBA; Rin1; RIT2; ROC2
Gene Symbols RIN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human RIN1 (NP_004283.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207119, 225870
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.