Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545810 Supplier Invitrogen™ Supplier No. PA545810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RTDVNLENGL EPAETHSMVR HKDGGYSEEE DVKTCARDSG YDSLSNRLSI Sequence homology: Cow: 80%; Dog: 80%; Horse: 80%; Human: 100%; Mouse: 83%; Pig: 80%; Rat: 83%.

The RAB5 protein is a small GTPase involved in membrane trafficking in the early endocytic pathway. The protein encoded by this gene binds the GTP-bound form of the RAB5 protein preferentially over the GDP-bound form, and functions as a guanine nucleotide exchange factor for RAB5. The encoded protein is found primarily as a tetramer in the cytoplasm and does not bind other members of the RAB family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIN2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RIN2
Gene Accession No. Q8WYP3
Gene Alias 2010003K16Rik; 4632403N06Rik; AW821980; MACS; RAB5 interacting protein 2; ras and Rab interactor 2; RAS association (RalGDS/AF-6) domain containing protein JC265; RAS association domain family 4; Ras inhibitor JC265; ras interaction/interference protein 2; RASSF4; Rin2
Gene Symbols RIN2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RIN2 (aa 37-86).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54453
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.