Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ring Finger Protein 213 (RNF213) Rabbit anti-Mouse, Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABC139125UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC1391-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC1391-25UG Supplier MilliporeSigma Supplier No. ABC139125UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Ring Finger Protein 213 (RNF213), Cat. No. ABC1391, is a rabbit polyclonal antibody that detects E3 ubiquitin-protein ligase RNF213 and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ring Finger Protein 213 (RNF213)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide.
Gene Alias E3 ubiquitin-protein ligase RNF213;ALK lymphoma oligomerization partner on chromosome 17;Mysterin;RING finger protein 213;RING-type E3 ubiquitin transferase RNF213
Gene Symbols RNF213;ALO17;C17orf27;KIAA1554;KIAA1618;MYSTR
Host Species Rabbit
Immunogen 11 GST-tagged recombinant fragments from human Ring Finger Protein 213.
Molecular Weight of Antigen ∽596 kDa observed; 591.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein A purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects human and mouse Ring Finger Protein 213.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.