Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RING1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204111AF750
Description
RING1 Polyclonal antibody specifically detects RING1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
RING1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
EC 6.3.2, EC 6.3.2.-, Polycomb complex protein RING1, Really interesting new gene 1 protein, ring finger protein 1RING1A, RNF1E3 ubiquitin-protein ligase RING1 | |
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human Ring Finger Protein 1 using the numbering given in entry NP_002922.2 (GeneID 6015). | |
0.1 mL | |
Zinc Finger | |
6015 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction