Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RING1 Monoclonal Antibody (8C12F4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531893
Description
MA5-31893 has been tested in indirect ELISA.
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4.
Specifications
RING1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q06587 | |
RING1 | |
Purified recombinant fragment of human Ring1 (AA: 79-263) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
8C12F4 | |
Unconjugated | |
RING1 | |
E3 ubiquitin-protein ligase RING1; Polycomb complex protein RING1; really interesting new gene 1 protein; RING finger protein 1; RING1; Ring1A; RING-type E3 ubiquitin transferase RING1; Rnf1; Transcription repressor Ring1A | |
Mouse | |
Protein G | |
RUO | |
6015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction