Learn More
RING1 Mouse anti-Human, Clone: OTI2D11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809239

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4.Specifications
RING1 | |
Monoclonal | |
Unconjugated | |
RING1 | |
RING1A, RNF1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6015 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2D11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q06587 | |
RING1 | |
Human recombit protein fragment corresponding to amino acids 88-362 of human RING1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.