Description
- The nitrile dipped palm and waterproof barrier combines protection against impact hazards with 360 cut resistance and liquid protection for a wide range of applications
- A breathable knit shell developed with Ringers' F3 Technology™
- Patented TPR design provide superior dexterity and comfort
- The waterproof liner helps keep hands dry while maintaining excellent dexterity
Specifications
Specifications
| Coating Material | Nitrile |
| Color | Gray |
| Cut Resistance Rating | ANSI A6 |
| Latex Free | Yes |
| Material | HPPE |
| Size | 9 |
| Style | Ringers™ R080 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.